Cyclopentylacetyl chloride ≥97%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 97%
CAS No.: 1122-99-2
Molecular Formula: C7H11ClO
Formula Weight: 146.62
Flash Point: 71°(160°F)
Refractive Index: 1.4625
Odour: Pungent
Boiling Point: 186°
Beilstein Reference: 1906113
UN No.: 3265
MDL No.: MFCD00236031
Formule:
C₇H₁₁ClO Kookpunt: 186 °C Dichtheid: 1,087 Vlampunt: 71 °C (160 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00236031 CAS nummer: 1122-99-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...