Acetonyltriphenylphosphonium bromide ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B22434.06
B22434.14
B22434.06EA
44.6
EUR
B22434.06
B22434.14
Acetonyltriphenylphosphonium bromide ≥98%
Acetonyltriphenylphosphonium bromide
Purity: 98+%
CAS No.: 2236-01-3
Molecular Formula: C21H20BrOP
Formula Weight: 399.27
Melting Point: 226-227°
Beilstein Reference: 3802148
Formule:
C₂₁H₂₀BrOP Smeltpunt: 226…227 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00051863 CAS nummer: 2236-01-3 EINECS: 218-796-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...