2,5-Dihydro-2,5-dimethoxy-2-methylfuran
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01757DA SB01757EA SB01757.50
MAYBSB01757DAEA 0 EUR
MAYBSB01757DA MAYBSB01757EA MAYBSB01757.50
2,5-Dihydro-2,5-dimethoxy-2-methylfuran
2,5-Dihydro-2,5-dimethoxy-2-methylfuran
Formule: C₇H₁₂O₃
MDL Nummer: MFCD00126939
CAS nummer: 22414-24-0
EINECS: 244-972-6