Carbenicillin disodium salt
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
SERA15875.03EA
0
EUR
SERA15875.03
SERA15875.01
SERA15875.02
Carbenicillin disodium salt
Carbenicillin disodium salt
Formule:
C₁₇H₁₆N₂O₆S·2Na MW: 422,363 g/mol Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00077683 CAS nummer: 4800-94-6 EINECS: 225-360-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...