(±)-Tetrahydro-2-furoic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Tetrahydrofuran-2-carboxylic acid, 2-Tetrahydrofuran Carboxylic Acid

CC16901.1 CC16901.10
MAYBCC16901.1EA 0 EUR
MAYBCC16901.1 MAYBCC16901.10
(±)-Tetrahydro-2-furoic acid
(±)-Tetrahydro-2-furoic acid
Formule: C₅H₈O₃
MDL Nummer: MFCD00022473
CAS nummer: 16874-33-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR