N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4058964.0001 4058964.0025
BACE4058964.0001EA 0 EUR
BACE4058964.0001 BACE4058964.0025
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
Formule: C₁₆H₃₃NO₆
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 85261-19-4