N-Cbz-glycine(Z-Gly-OH)

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

4000228.0100 4000228.0500
BACE4000228.0100EA 0 EUR
BACE4000228.0100 BACE4000228.0500
N-Cbz-glycine(Z-Gly-OH)
N-Cbz-glycine(Z-Gly-OH)
Formule: C₁₀H₁₁NO₄
MDL Nummer: MFCD00002691
CAS nummer: 1138-80-3
EINECS: 214-516-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR