Naphthol AS-BI phosphate
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 7-Bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, Naphthol AS-BI Phosphoric acid solution, Naphthol AS-BI-Phosphate, 6-Bromo-3-((2-methoxyphenyl)carbamoyl)naphthalen-2-yl dihydrogen phosphate
29988.01 29988.02 29988.03 29988.04
SERA29988.01EA 0 EUR
SERA29988.01 SERA29988.02 SERA29988.03 SERA29988.04
Naphthol AS-BI phosphate
Naphthol AS-BI phosphate
Formule: C₁₈H₁₅BrNO₆P
MW: 452.1926 g/mol
MDL Nummer: MFCD00004058
CAS nummer: 1919-91-1
EINECS: 217-645-0