Guanosine-5'-diphosphate disodium salt
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24291.02
24291.03
SERA24291.02EA
0
EUR
SERA24291.02
SERA24291.03
Guanosine-5'-diphosphate disodium salt
Guanosine-5'-diphosphate disodium salt
Formule:
C₁₀H₁₃N₅Na₂O₁₁P₂ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00070121 CAS nummer: 7415-69-2 EINECS: 231-026-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...