n-Dodecyl-β-D-maltoside 99%, high purity
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: DDM
329370010 329370050 329370250
ACRO329370010EA 191 EUR
ACRO329370010 ACRO329370050 ACRO329370250
n-Dodecyl-β-D-maltoside 99%, high purity
n-Dodecyl-β-D-maltoside
Formule: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
Smeltpunt: 217 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00043012
CAS nummer: 69227-93-6
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥98.5 % (ELSD)
Specific optical rotation +45° to +49.5° (20°C, 589 nm) (c=1, water)