n-Dodecyl-β-D-maltoside
Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: DDM
4061027.0001 4061027.0005 4061027.0025
BACE4061027.0001EA 0 EUR
BACE4061027.0001 BACE4061027.0005 BACE4061027.0025
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
Formule: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00043012
CAS nummer: 69227-93-6