n-Octyl-β-D-maltoside
Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
54560-1 54560-2
BMOL54560-1EA 0 EUR
BMOL54560-1 BMOL54560-2
n-Octyl-β-D-maltoside
n-Octyl-β-D-maltoside
Formule: C₂₀H₃₈O₁₁
CAS nummer: 82494-08-4