DNA (Deoxyribonucleic acid) (from fish sperm)
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18580.01 18580.02
SERA18580.01EA 76 EUR
SERA18580.01 SERA18580.02
DNA (Deoxyribonucleic acid) (from fish sperm)
DNA (Deoxyribonucleic acid)
MDL Nummer: MFCD00130922
CAS nummer: 9007-49-2
EINECS: 309-566-6