Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4006587.0050 4006587.1000
BACE4006587.0050EA 0 EUR
BACE4006587.0050 BACE4006587.1000
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Eiwitten en peptiden

  • EINECS: 261-529-2

Order Now

Specificaties

Protein/peptide name Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Environmentally Preferable
Protein synonyms Ubenimex|Bestatin-S-R-S-N-3-amino-2-hydroxy-4-phenylbutyroyl-L-leucine|N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine
Species
Bron
CAS No. 58970-76-6
MDL No. MFCD00083262
Moleculair gewicht 308,4 kDa
Molecular formula C₁₆H₂₄N₂O₄
Storage conditions Freezer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR