3,3'-Diaminobenzidine tetrahydrochloride dihydrate, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
32750-1G-F 32750-25G-F 32750-5G-F
32750-1G-FEA 0 EUR
32750-1G-F 32750-25G-F 32750-5G-F
3,3'-Diaminobenzidine tetrahydrochloride dihydrate, Supelco®
3,3'-Diaminobenzidine tetrahydrochloride dihydrate
Formule: C₁₂H₁₈Cl₄N₄·2H₂O
CAS nummer: 167684-17-5
EINECS: 231-018-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=985681e0897fbc47b026a09a6c22b2cb0b881ef2&vid=7d041fc8aff47ea2e7611dc25645c3f41b544944&prid=web&clid=SAPDAM
Order Now
Specificaties
Environmentally Preferable