Fluorescent inks, Blak-Ray®
Leverancier: UVP ULTRA VIOLET PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
96-0021-04 96-0021-05 96-0021-01 96-0021-02 96-0057-04
UVPA96-0021-04EA 435 EUR
UVPA96-0021-04 UVPA96-0021-05 UVPA96-0021-01 UVPA96-0021-02 UVPA96-0057-04
Fluorescent inks, Blak-Ray®
Inkten
Blak-Ray® fluorescent inks are suited for use on skin, fabric and similar porous or textured surfaces.
  • Simple to use - inks come ready made
  • Won't gum up - stamp pads stay cleaner longer
  • Economical - small amounts of ink are used with stamp pads
  • All inks are formulated to fluoresce brightly under UVP's longwave ultraviolet lamps
  • Readmission type inks are designed for use at amusement parks, social gatherings and sporting events
  • Inks are non-toxic, fast-drying and non-transferable
Suitable for readmission control, inspection, coding and inventory control.