Adapters, tubing, KA, KECK™, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286345105 286345002 286345208 286345302
271-7152EA 7.2 EUR
271-7152 271-7151 271-7155 271-7156
Adapters, tubing, KA, KECK™, DURAN®
Fittingen en connectoren
KECK™ adapter for glass thread KA with screw threaded cap.
  • Connect or remove hose by twisting 1/4 turn
  • Available for sizes GL 14, RD 14 and SQ 13
It protects the thread from damage and guarantee fast, safe and leakproof (upto 30 bar pressure and 0,66 bar vacuum) connections.
Leveringsinformatie: The adapters are available as a complete set which comes either with 4,5 / 9,0 mm Ø tubing part.