Infusion filter, Intrapur® lipid
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRAU4099702EA 1730 EUR
BRAU4099702
Infusion filter, Intrapur® lipid
Infusen
Intrapur Lipid is a 1,2 μm infusion filter for lipid emulsions and all-in-one parenteral nutrition.
  • Retention of fungi, spores and particles
  • Reliable air venting regardless of position
  • High throughput
  • Pressure resistant to 3,1 bar
  • Luer lock fitting