Sleeves for ground joints
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42388010 42388019 42388024 42388029
HECH42388010EA 23 EUR
HECH42388010 HECH42388019 HECH42388024 HECH42388029
Sleeves for ground joints
Geslepen Geslepen hulzen
Disposable sleeves for ground joints and stoppers, prevent the ground joints from sticking together.
  • Made of PTFE
  • Self-lubricating
  • Outstanding chemical resistance at temperatures