Injection spikes for multidose vials, Mini Spike Plus®
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4550234 4550340 4550560 4550510 4550242 4550528 4550587 4550579
612-0193EA 464 EUR
612-0193 612-0195 BRAU4550560 BRAU4550510 BRAU4550242 BRAU4550528 BRAU4550587 BRAU4550579
Injection spikes for multidose vials, Mini Spike Plus®
Infusen
Mini Spike Plus withdrawal and injection spikes for multidose vials and multiple withdrawals.
  • Bacteria-proof venting and comfortable handling
  • Higher safety, increased efficiency, high flow rate
  • Effortless and precise penetration even of thick stoppers
Integrated snap cap suitable for one-handed opening and closing with a reliable protection of drugs against microbial contamination during withdrawal.