Nuts, hexagonal, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H1132-26 H1132-22 H1132-18 H1132-14
BOHLH1132-26EA 18.3 EUR
BOHLH1132-26 BOHLH1132-22 BOHLH1132-18 BOHLH1132-14
Nuts, hexagonal, BOLA
Bevestigingen Moeren
PTFE, white.
  • Temperature resistance: from -200 °C to +250 °C
  • Universal chemical resistance
  • Similar to DIN 934 / DIN EN ISO 4032