Mixing measuring cylinders, tall form, class B, soda-lime glass
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42211018 42211024 42211048 42211060
612-9440EA 15.9 EUR
612-9440 612-9441 612-9443 612-9444
Mixing measuring cylinders, tall form, class B, soda-lime glass
Cilinders Maatcilinders
Soda glass, class B, with PE stopper.
  • Calibrated to contain (TC, In)
  • Fused amber graduation
  • Interchangeable base and protecting ring of HDPE
Certificaten: DIN EN ISO 4788