Measuring cylinders, tall form, class B, borosilicate glass
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42180030 42180024 42180060 42180066
HECH42180030EA 10.4 EUR
HECH42180030 HECH42180024 HECH42180060 HECH42180066
Measuring cylinders, tall form, class B, borosilicate glass
Cilinders Maatcilinders
Borosilicate glass, tall form, class B.
  • Calibrated to contain 'ln'
  • Hexagonal glass base, with spout
  • Fused brown graduations
Certificaten: DIN EN ISO 4788