Sulfonation flasks, with four necks
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0.850.050 0.850.035 0.850.031 0.850.032 0.850.074 0.850.101 0.850.600
WITG0.850.050EA 159 EUR
WITG0.850.050 WITG0.850.035 WITG0.850.031 WITG0.850.032 WITG0.850.074 WITG0.850.101 WITG0.850.600
Sulfonation flasks, with four necks
Kolven Kolven voor sulfonering
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two parallel and two angled side necks.
Order Now
Specificaties
Environmentally Preferable