Reaction vessels, cylindrical
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0.996.001 0.996.002 0.995.500
WITG0.996.001EA 1250 EUR
WITG0.996.001 WITG0.996.002 WITG0.995.500
Reaction vessels, cylindrical
Reactievaatjes Reactievaten, cilindrische vorm
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat bottom, jacketed, flange with groove and draining valve.