Distilling heads
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.010.006 1.010.001 1.010.004 1.010.005
201-0207EA 33.9 EUR
201-0207 WITG1.010.001 WITG1.010.004 WITG1.010.005
Distilling heads
Onderdelen voor destillatie Destillatiekoppen
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With one NS socket and two NS cones, descending angle: 75°.