Distilling bridges
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.200.300 1.200.200
WITG1.200.300EA 58.9 EUR
WITG1.200.300 WITG1.200.200
Distilling bridges
Onderdelen voor destillatie Destillatiebruggen
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling links without condenser, with two vertical NS cones.