Volumetric flasks, class A, blue markings
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
92209800105 92209800205 92209800409 92209800509 92209800810 42096008 92209800303
HECH92209800105EA 147 EUR
HECH92209800105 HECH92209800205 HECH92209800409 HECH92209800509 HECH92209800810 HECH42096008 HECH92209800303
Volumetric flasks, class A, blue markings
Kolven Maatkolven
Borosilicate glass 3.3.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in blue
  • Conformity certified (DE-M marking)
Certificaten: DIN EN ISO 1042