Condensers, Davies
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.739.163 1.739.253
WITG1.739.163EA 108 EUR
WITG1.739.163 WITG1.739.253
Condensers, Davies
Condensors
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two glass olives.