Dropping funnels, spherical, without pressure balance
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.892.100 1.892.250 1.892.500
WITG1.892.100EA 61.4 EUR
WITG1.892.100 WITG1.892.250 WITG1.892.500
Dropping funnels, spherical, without pressure balance
Trechters Druppeltrechters
Dropping funnels, spherical, made of borosilicate glass 3.3, stopcock with glass key. With NS cone and socket.
  • Without graduations
  • Excellent chemical resistance
  • Excellent resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes