Anatomical Charts
Leverancier: 3B Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1001530 1001522 1001470 1001468 1001496 1001480 1001494 1001500 1001524 1001510 1001528 1001586
BINH1001530EA 0 EUR
BINH1001530 761-0473 761-0467 761-0466 761-0470 761-0468 761-0469 761-0471 761-0474 761-0472 761-0475 761-0476
Anatomical Charts
Kaarten
These colourful anatomical charts depict various parts of the human body.
  • Thick lamination ensures the chart does not curl up at the edges
  • UV treatment ensures the chart does not get a faded yellow colour over time
  • Printed on premium glossy UV resistant paper and thickly laminated on both sides
Leveringsinformatie: Supplied with metal eyelets for easy hanging.