Micro soxhlet extraction apparatus, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
415500 415502 415501 W416788
WHEA415500EA 684 EUR
WHEA415500 WHEA415502 WHEA415501 WHEAW416788
Micro soxhlet extraction apparatus, WHEATON®
Extractors Soxhlet-extractors en Soxhlet-type extractors
Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.