Fractionating columns, Hempel, with glass jacket
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.503.500 1.503.300
WITG1.503.500EA 72.3 EUR
WITG1.503.500 WITG1.503.300
Fractionating columns, Hempel, with glass jacket
Onderdelen voor destillatie Destillatiekolommen
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For packings like Raschig rings or glass beads, with NS socket and NS cone, with fixed vacuum jacket.