Drying tubes, U-shaped
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2.114.100 2.114.125 2.114.150 2.114.250
WITG2.114.100EA 52.1 EUR
WITG2.114.100 WITG2.114.125 WITG2.114.150 WITG2.114.250
Drying tubes, U-shaped
Buisjes Droogbuizen
Borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes