Graduated pipettes, Class AS, total delivery, FORTUNA®
Leverancier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.11425404 1.11421404 1.11423404 1.11443404 1.11447404 1.11421402 1.11423402 1.11427402 1.11441404 1.11419402 1.11425402 1.11437404 1.11447402
POUL1.11425404EA 18.5 EUR
POUL1.11425404 POUL1.11421404 POUL1.11423404 POUL1.11443404 POUL1.11447404 POUL1.11421402 POUL1.11423402 POUL1.11427402 POUL1.11441404 POUL1.11419402 POUL1.11425402 POUL1.11437404 POUL1.11447402
Graduated pipettes, Class AS, total delivery, FORTUNA®
Pipetten Gegradueerde pipetten
Soda-lime glass.
  • Blue or amber graduations
  • Ring marks
  • Batch certificate
EN ISO 835:2007. Conformity approved.