Graduated pipettes, with piston, FORTUNA®
Leverancier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.1424702 1.1423204
612-1007EA 93.5 EUR
612-1007 612-1505
Graduated pipettes, with piston, FORTUNA®
Pipetten Gegradueerde pipetten
Clear glass.
  • Blue or amber graduations
  • Clear calibrated cylinders
  • Tolerance indications
  • Use of pipetting aid not necessary
  • Single pack
Without certificate.