Stand bases
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5031 5030 5032 5033 5034 5037 5038 5035 5036
BOCH5031EA 41.3 EUR
BOCH5031 BOCH5030 BOCH5032 241-3167 BOCH5034 BOCH5037 BOCH5038 BOCH5035 BOCH5036
Stand bases
Staafstatieven
Steel, varnished.
  • With M10 threaded hole
  • One adjustable foot
Certificaten: DIN 12892
Order Now
Specificaties
Environmentally Preferable