Cup samplers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5030A-1500 5030A-1000 5030A-600 5030J-1000 5030J-1500
SAMP5030A-1500EA 731 EUR
SAMP5030A-1500 SAMP5030A-1000 SAMP5030A-600 SAMP5030J-1000 SAMP5030J-1500
Cup samplers
Monsternemers Monsternemers voor vloeistoffen
Stainless steel cup samplers designed for liquids and suspensions.
  • Single point liquid sampler with removable cup
  • Simple operation, easy to clean and hygienic design
  • Discrete samples to be taken from known heights within the product