Jars, Wide Mouth, with or without Screw Cap, Rixius
Leverancier: RIXIUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4-10011500130B 41001022565WC 41001050090WC 41001065090WC 410011000WC
RIXI4-10011500130BEA 184 EUR
RIXI4-10011500130B RIXI41001022565WC RIXI41001050090WC RIXI41001065090WC RIXI410011000WC
Jars, Wide Mouth, with or without Screw Cap, Rixius
Potjes Potjes voor algemeen gebruik
PP, white.
  • Good chemical resistance
  • Good temperature resistance
Ideal for sampling, transportation and storage.