Fluid samplers, Liquid Master
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5035S-1000 5035J-1500 5035S-1500 5035S-600 5035J-1000 5035J-2000 5035S-2000
SAMP5035S-1000EA 664 EUR
SAMP5035S-1000 SAMP5035J-1500 SAMP5035S-1500 SAMP5035S-600 SAMP5035J-1000 SAMP5035J-2000 SAMP5035S-2000
Fluid samplers, Liquid Master
Monsternemers Monsternemers voor vloeistoffen
The Liquid Master has been designed to be quickly stripped down for cleaning.
  • Quick and easy to use
  • Push button to press to take samples
  • For a larger volume, the sampler can be inserted deep into the product
Suitable for taking samples from low and medium viscosity liquids.