Measuring cylinders, tall form, class A, borosilicate glass, FORTUNA®
Leverancier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.3103302 1.3103702 1.3103704
POUL1.3103302EA 24.1 EUR
POUL1.3103302 POUL1.3103702 POUL1.3103704
Measuring cylinders, tall form, class A, borosilicate glass, FORTUNA®
Cilinders Maatcilinders
Borosilicate glass 3,3 tall form, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, with spout
  • Available with blue or brown graduations
Certificaten: DIN EN ISO 4788