Volumetric flasks, class A, glass, clear, GL screw cap
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.642.010 3.642.100 3.642.003 3.642.025 3.642.005 3.642.050
WITG3.642.010EA 26.2 EUR
WITG3.642.010 WITG3.642.100 WITG3.642.003 WITG3.642.025 WITG3.642.005 WITG3.642.050
Volumetric flasks, class A, glass, clear, GL screw cap
Kolven Maatkolven
Borosilicate glass 3.3 brown markings.
  • DE-M marked according to the German calibration law
  • Iinclusive batch certificate
  • With GL thread and screw cap
Certificaten: Tolerance according to DIN EN ISO 1042