Graduated pipettes for encymatic analysis, Class AS
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.517.011 3.517.710 3.517.021 3.517.505 3.517.051 3.517.101 3.517.201 3.517.202
WITG3.517.011EA 42.8 EUR
WITG3.517.011 WITG3.517.710 WITG3.517.021 WITG3.517.505 WITG3.517.051 WITG3.517.101 WITG3.517.201 WITG3.517.202
Graduated pipettes for encymatic analysis, Class AS
Pipetten Gegradueerde pipetten
Pipettes for enzymatic analysis.
  • For partial delivery
  • Main point ring-graduation
  • Diffico-brown
  • Calibrated to contain „EX"
DIN 12699
Certificaten: DIN 12699. Conformity certified (except for article with *).