Graduated pipettes, Class AS
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.510.021 3.510.022
613-2102EA 49.9 EUR
613-2102 613-2103
Graduated pipettes, Class AS
Pipetten Gegradueerde pipetten
Graduated pipettes, class AS, conformity certified.
  • For complete swift delivery
  • With amber stain main point ring-graduation
  • Tempered delivery tips offering a wear resistance
  • With construction in the suction to hold cotton plug
  • Waiting time 3 s
Certificaten: DIN 12697