Graduated pipettes, with piston, Class B
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.575.101 3.575.505 3.575.710 3.575.910 3.575.051
613-2170EA 15 EUR
613-2170 613-2171 613-2172 613-2173 WITG3.575.051
Graduated pipettes, with piston, Class B
Pipetten Gegradueerde pipetten
Glass, class DIN-B
  • With ISO-COLOR-Code
  • With interchangeable piston and tempered tips
  • With amber stain DIFFICO graduation and inscriptions
  • Short line graduation
  • Complete delivery