Burettes, lateral stopcock, class AS, Schellbach
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.012.905 3.012.255 3.012.505
661-4120EA 65.9 EUR
661-4120 661-4121 661-4122
Burettes, lateral stopcock, class AS, Schellbach
Buretten
Burettes, class AS, Schellbach blue line, conformity certified and made of clear borosilicate glass.
  • Main point ring graduation
  • Lateral stopcock, with PTFE-needle valve
  • Overtwist device