Separating funnels, Squibb
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2.649.125 2.649.150
WITG2.649.125EA 65.9 EUR
WITG2.649.125 WITG2.649.150
Separating funnels, Squibb
Trechters Scheitrechters
Separatory funnels, Squibb, made of borosilicate glass 3.3, stopcock with economy PTFE-needle valve.
  • Easy to read graduation
  • Very good chemical resistance
  • Very good temperature resistance