Semi-solid samplers, MaxiCorer
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7010A-01 7010A-02
SAMP7010A-01EA 175 EUR
SAMP7010A-01 SAMP7010A-02
Semi-solid samplers, MaxiCorer
Monsternemers Monsternemers voor vaste stoffen
The MaxiCorer can be used for sampling or dispensing small quantities of products. This designed for powders and granules, as well as for semi-solids such as cheese and thick grease in production, quality control or laboratories.
  • Fast and efficent core sampler
  • Long and curved blade construction
  • Hygienic design that can be easily cleaned