Hand shovels for industry
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8300-1003 8300-1001
BURK8300-1003EA 33.2 EUR
BURK8300-1003 BURK8300-1001
Hand shovels for industry
Spaden en schoppen
With anti-static additive, ideal for robust use in chemistry and industry. Polypropylene has a high resistance to chemicals and temperatures.
  • Anti-static
  • Unbreakable
  • Easy to clean
  • Light-weight
  • Autoclavable at 121 °C