Bottles, narrow neck, square, HDPE, graduated, with screw caps
Leverancier: LP Italiana
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LPIT294232EA 142 EUR
LPIT294232
Bottles, narrow neck, square, HDPE, graduated, with screw caps
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Graduated
Screw cap with gasket.